Tris(2-aminoethyl)amine, 96%, Thermo Scientific Chemicals
Tris(2-aminoethyl)amine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris(2-aminoethyl)amine, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 314791000
Price (USD)/ Each
190.65
Online exclusive
212.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
190.65
Online exclusive
212.00 
Save 21.35 (10%)
Add to cart
Tris(2-aminoethyl)amine, 96%, Thermo Scientific Chemicals
Catalog number314791000
Price (USD)/ Each
190.65
Online exclusive
212.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS4097-89-6
IUPAC Nametris(2-aminoethyl)amine
Molecular FormulaC6H18N4
InChI KeyMBYLVOKEDDQJDY-UHFFFAOYSA-N
SMILESNCCN(CCN)CCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4960 to 1.4980 (20°C, 589 nm)
GC>=98.0 %
Appearance (Color)Clear colorless to yellow-green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only