N,N'-Dimethyl-1,3-propanediamine, 97%, Thermo Scientific Chemicals
N,N'-Dimethyl-1,3-propanediamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N'-Dimethyl-1,3-propanediamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 314830250
Price (USD)/ Each
221.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
221.00
Add to cart
N,N'-Dimethyl-1,3-propanediamine, 97%, Thermo Scientific Chemicals
Catalog number314830250
Price (USD)/ Each
221.00
-
Add to cart
Chemical Identifiers
CAS111-33-1
IUPAC Namemethyl[3-(methylazaniumyl)propyl]azanium
Molecular FormulaC5H16N2
InChI KeyUQUPIHHYKUEXQD-UHFFFAOYSA-P
SMILESC[NH2+]CCC[NH2+]C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.4370 to 1.4410 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only