Neodymium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals
Neodymium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Neodymium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 315070050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10024-93-8
IUPAC Nameneodymium(3+) trichloride
Molecular FormulaCl3Nd
InChI KeyATINCSYRHURBSP-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Nd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to light green
Neodymium (Nd)55.8 to 59.3 %
Total conc. of imp.as REO=<0.1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only