Thermo Scientific Chemicals

Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 315140250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Erbium(III) nitrate pentahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 315140250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 315140250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10031-51-3
IUPAC Name
erbium(3+) pentahydrate trinitrate
Molecular Formula
ErH10N3O14
InChI Key
LWHHUEHWVBVASY-UHFFFAOYSA-N
SMILES
O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
Pink
Appearance (Form)
Crystalline powder or crystals
Assay
33 to 39 % (Er)
Total conc. of imp.as REO
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text