Thermo Scientific Chemicals

Bismuth(III) nitrate pentahydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 315151000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Bismuth(III) nitrate pentahydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 315151000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 315151000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10035-06-0
IUPAC Name
bismuth(3+) pentahydrate trinitrate
Molecular Formula
BiH10N3O14
InChI Key
FBXVOTBTGXARNA-UHFFFAOYSA-N
SMILES
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
White
Total trace metal impurities
=<0.001 %
Appearance (Form)
Crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text