Thermo Scientific Chemicals

Strontium nitrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 315170025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Strontium nitrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 315170025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 315170025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10042-76-9
IUPAC Name
strontium(2+) dinitrate
Molecular Formula
N2O6Sr
InChI Key
DHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
White
Loss on drying
=<0.1 % (105°C)
Heavy metals (ICP-OES)
=<5 ppm
Chloride (Cl)
=<20 ppm
Sodium (Na)
=<1000 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text