Thermo Scientific Chemicals

Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals

Catalog number: 315181000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals

Catalog number: 315181000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 315181000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10043-35-3
IUPAC Name
boric acid
Molecular Formula
BH3O3
InChI Key
KGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILES
OB(O)O
Appearance (Form)
Crystalline powder or crystals
Titration with NaOH
>=98.5 %
Appearance (Color)
White
Total trace metal impurities
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text