Thermo Scientific Chemicals

Chromium(II) chloride, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 315190010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Chromium(II) chloride, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 315190010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 315190010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10049-05-5
IUPAC Name
λ²-chromium(2+) dichloride
Molecular Formula
Cl2Cr
InChI Key
XBWRJSSJWDOUSJ-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Cr++]
Appearance (Color)
Off-white to grey-green
Appearance (Form)
Powder
Total trace metal impurities
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text