Thermo Scientific Chemicals

Lanthanum chloride, 99.9%, Thermo Scientific Chemicals

Catalog number: 315230500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Lanthanum chloride, 99.9%, Thermo Scientific Chemicals

Catalog number: 315230500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 315230500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10099-58-8
IUPAC Name
lanthanum(3+) trichloride
Molecular Formula
Cl3La
InChI Key
ICAKDTKJOYSXGC-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[La+3]
Appearance (Form)
Powder
Total conc. of imp.as REO
=<0.1 %
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text