Lead(II) nitrate, 99%, for analysis
Lead(II) nitrate, 99%, for analysis
Lead(II) nitrate, 99%, for analysis
Lead(II) nitrate, 99%, for analysis
Thermo Scientific Chemicals

Lead(II) nitrate, 99%, for analysis

CAS: 10099-74-8 | N2O6Pb | 331.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 315242500
Price (USD)
32.65
Online Exclusive
36.30
Save 3.65 (10%)
Each
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
32.65
Online Exclusive
36.30
Save 3.65 (10%)
Each
Add to cart
Chemical Identifiers
CAS10099-74-8
IUPAC Nameλ²-lead(2+) dinitrate
Molecular FormulaN2O6Pb
InChI KeyRLJMLMKIBZAXJO-UHFFFAOYSA-N
SMILES[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Copper (Cu)=<0.001 %
Titration Complexometric>=98.5 %
Insoluble matter=<0.005 % (10 % in dilute HNO3)
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link