Thermo Scientific Chemicals

(1S,2R)-(-)-cis-1-Amino-2-indanol, 99%, Thermo Scientific Chemicals

Catalog number: 315650050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

(1S,2R)-(-)-cis-1-Amino-2-indanol, 99%, Thermo Scientific Chemicals

Catalog number: 315650050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 315650050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
126456-43-7
IUPAC Name
(1S,2R)-2-hydroxy-2,3-dihydro-1H-inden-1-aminium
Molecular Formula
C9H12NO
InChI Key
LOPKSXMQWBYUOI-BDAKNGLRSA-O
SMILES
[NH3+][C@@H]1[C@H](O)CC2=CC=CC=C12
Infrared spectrum
Conforms
Melting point
117°C to 121°C
Appearance (Form)
Powder
Specific optical rotation
-41.5° to -46.5° (20°C, 589 nm) (c=1, CH3OH)
Appearance (Color)
White to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text