Thermo Scientific Chemicals

Magnesium hydroxide, 95%, pure, Thermo Scientific Chemicals

Catalog number: 315720250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Magnesium hydroxide, 95%, pure, Thermo Scientific Chemicals

Catalog number: 315720250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 315720250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1309-42-8
IUPAC Name
magnesium(2+) dihydroxide
Molecular Formula
H2MgO2
InChI Key
VTHJTEIRLNZDEV-UHFFFAOYSA-L
SMILES
[OH-].[OH-].[Mg++]
Appearance (Color)
White
Appearance (Form)
Powder
Titration Complexometric
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text