Thermo Scientific Chemicals

Sodium peroxide, ACS reagent, Thermo Scientific Chemicals

Catalog number: 315760250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium peroxide, ACS reagent, Thermo Scientific Chemicals

Catalog number: 315760250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 315760250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1313-60-6
IUPAC Name
disodium dioxidanediide
Molecular Formula
Na2O2
InChI Key
PFUVRDFDKPNGAV-UHFFFAOYSA-N
SMILES
[Na+].[Na+].[O-][O-]
Appearance
May contain a small quantity of discolored
Phosphate (PO4)
=<5 ppm
Titration with KMnO4
>=93.0 %
Heavy metals (as Pb)
=<20 ppm
Sulfate (SO4)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text