Thermo Scientific Chemicals

Phosphorus pentoxide, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 315820010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Phosphorus pentoxide, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 315820010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 315820010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-56-3
IUPAC Name
tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
Molecular Formula
O5P2
InChI Key
DLYUQMMRRRQYAE-UHFFFAOYSA-N
SMILES
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
Heavy metals (as Pb)
=<100 ppm
Phosphorus trioxide (P2O3)
Passes test (limit about 0.02 %)
Appearance (Color)
White
Appearance (Form)
Powder
Titration with NaOH
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text