Lead(II) oxide, 99.9+%, (trace metal basis), <10 microns, powder, Thermo Scientific Chemicals
Lead(II) oxide, 99.9+%, (trace metal basis), &lt;10 microns, powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) oxide, 99.9+%, (trace metal basis), <10 microns, powder, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Glass bottle
Catalog number 315850025
Price (USD)/ Each
308.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
363.00 
Save 54.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
308.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
363.00 
Save 54.35 (15%)
Add to cart
Lead(II) oxide, 99.9+%, (trace metal basis), <10 microns, powder, Thermo Scientific Chemicals
Catalog number315850025
Price (USD)/ Each
308.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
363.00 
Save 54.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1317-36-8
IUPAC Nameplumbanone
Molecular FormulaOPb
InChI KeyYEXPOXQUZXUXJW-UHFFFAOYSA-N
SMILESO=[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to light orange
Average particle size=<10 µm
Total trace metal impurities=<1000.0 ppm
Appearance (Form)Powder

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For ultra trace analysis
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only