Thermo Scientific Chemicals

Lead(II) oxide, 99.9+%, (trace metal basis), <10 microns, powder, Thermo Scientific Chemicals

Catalog number: 315855000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Lead(II) oxide, 99.9+%, (trace metal basis), <10 microns, powder, Thermo Scientific Chemicals

Catalog number: 315855000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 315855000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1317-36-8
IUPAC Name
plumbanone
Molecular Formula
OPb
InChI Key
YEXPOXQUZXUXJW-UHFFFAOYSA-N
SMILES
O=[Pb]
Appearance (Color)
Yellow to light orange
Average particle size
=<10 µm
Total trace metal impurities
=<1000.0 ppm
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For ultra trace analysis
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text