Thermo Scientific Chemicals

Iron(II) sulfide, 99.9%, (trace metal basis), -100 mesh, Thermo Scientific Chemicals

Catalog number: 315870100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Iron(II) sulfide, 99.9%, (trace metal basis), -100 mesh, Thermo Scientific Chemicals

Catalog number: 315870100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 315870100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1317-37-9
IUPAC Name
λ²-iron(2+) sulfanediide
Molecular Formula
FeS
InChI Key
GNVXPFBEZCSHQZ-UHFFFAOYSA-N
SMILES
[S--].[Fe++]
Total trace metal impurities
=<0.1 %
Appearance (Color)
Grey to brown to black
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text