Thermo Scientific Chemicals

Cesium fluoride, 99.9%, extra pure, Thermo Scientific Chemicals

Catalog number: 315910050
5 g, Plastic bottle, Each
Thermo Scientific Chemicals

Cesium fluoride, 99.9%, extra pure, Thermo Scientific Chemicals

Catalog number: 315910050
5 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 315910050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13400-13-0
IUPAC Name
caesium(1+) fluoride
Molecular Formula
CsF
InChI Key
XJHCXCQVJFPJIK-UHFFFAOYSA-M
SMILES
[F-].[Cs+]
Appearance (Color)
White
Total trace metal impurities
=<0.1 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text