Thermo Scientific Chemicals

Potassium tetrachloroaurate(III), 98%, Thermo Scientific Chemicals

Catalog number: 316120010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Potassium tetrachloroaurate(III), 98%, Thermo Scientific Chemicals

Catalog number: 316120010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 316120010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13682-61-6
IUPAC Name
gold(3+) potassium tetrachloride
Molecular Formula
AuCl4K
InChI Key
JMONCQYBAMKVQQ-UHFFFAOYSA-J
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[K+].[Au+3]
Appearance (Form)
Crystalline powder
Appearance (Color)
Yellow to light orange
Assay
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text