Samarium(III) nitrate hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
Samarium(III) nitrate hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Samarium(III) nitrate hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 316151000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS13759-83-6
IUPAC Namesamarium(3+) hexahydrate trinitrate
Molecular FormulaH12N3O15Sm
InChI KeyHDCOFJGRHQAIPE-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Samarium (Sm)33.16 to 34.54 %
Appearance (Form)Crystalline powder or crystals
Total conc. of imp.as REO=<0.1 %
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only