Thermo Scientific Chemicals

Sodium hexachloroplatinate(IV) hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 316750010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium hexachloroplatinate(IV) hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 316750010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 316750010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
19583-77-8
IUPAC Name
disodium hexachloroplatinum hexahydrate
Molecular Formula
Cl6H12Na2O6Pt
InChI Key
XGFSWVNCHVSCSP-UHFFFAOYSA-H
SMILES
O.O.O.O.O.O.[Na+].[Na+].Cl[Pt](Cl)(Cl)(Cl)(Cl)Cl
Assay
>=97.5 %
Appearance (Form)
Crystalline solid
Appearance (Color)
Orange

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text