Thermo Scientific Chemicals

Bis(2-diphenylphosphinoethyl)phenylphosphine, 97%, Thermo Scientific Chemicals

Catalog number: 316820010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Bis(2-diphenylphosphinoethyl)phenylphosphine, 97%, Thermo Scientific Chemicals

Catalog number: 316820010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 316820010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
23582-02-7
IUPAC Name
(2-{[2-(diphenylphosphanyl)ethyl](phenyl)phosphanyl}ethyl)diphenylphosphane
Molecular Formula
C34H33P3
InChI Key
AXVOAMVQOCBPQT-UHFFFAOYSA-N
SMILES
C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to off-white
Melting point
126°C to 132°C
Infrared spectrum
Conforms
Appearance (Form)
Powder and/or chunks
Elemental Analysis
74.1 to 78.7 % (C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text