Thermo Scientific Chemicals

Tris[2-(diphenylphosphino)ethyl]phosphine, 97%, Thermo Scientific Chemicals

Catalog number: 316830050
5 g, Each
Thermo Scientific Chemicals

Tris[2-(diphenylphosphino)ethyl]phosphine, 97%, Thermo Scientific Chemicals

Catalog number: 316830050
5 g, Each
Quantity
Catalog number: 316830050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
23582-03-8
IUPAC Name
(2-{bis[2-(diphenylphosphanyl)ethyl]phosphanyl}ethyl)diphenylphosphane
Molecular Formula
C42H42P4
InChI Key
TVLNGWSWPKIYAO-UHFFFAOYSA-N
SMILES
C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Infrared spectrum
Conforms
Melting point
134°C to 139°C
Appearance (Form)
Powder
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text