Phosphomolybdic acid, ammonium salt hydrate, Thermo Scientific Chemicals
Phosphomolybdic acid, ammonium salt hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phosphomolybdic acid, ammonium salt hydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Packaging:
Plastic bottle
Catalog number 317100010
Price (USD)/ Each
1,019.65
Online exclusive
1133.00 
Save 113.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
1,019.65
Online exclusive
1133.00 
Save 113.35 (10%)
Add to cart
Phosphomolybdic acid, ammonium salt hydrate, Thermo Scientific Chemicals
Catalog number317100010
Price (USD)/ Each
1,019.65
Online exclusive
1133.00 
Save 113.35 (10%)
-
Add to cart
Chemical Identifiers
CAS54723-94-3
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<50 ppm
Appearance (Color)Yellow
Insoluble matter=<0.01 %
Appearance (Form)Powder
Calcium (Ca)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only