Thermo Scientific Chemicals

Phosphomolybdic acid, ammonium salt hydrate, Thermo Scientific Chemicals

Catalog number: 317100500
50 g, Plastic bottle, Each
Thermo Scientific Chemicals

Phosphomolybdic acid, ammonium salt hydrate, Thermo Scientific Chemicals

Catalog number: 317100500
50 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 317100500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
54723-94-3
Iron (Fe)
=<50 ppm
Appearance (Color)
Yellow
Insoluble matter
=<0.01 %
Appearance (Form)
Powder
Calcium (Ca)
=<200 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text