Thermo Scientific Chemicals

Triphenyltin chloride, 95%, Thermo Scientific Chemicals

Catalog number: 317290050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Triphenyltin chloride, 95%, Thermo Scientific Chemicals

Catalog number: 317290050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 317290050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
639-58-7
IUPAC Name
triphenyltin chloride
Molecular Formula
C18H17ClSn
InChI Key
ZDYYCRIXXMWWQC-UHFFFAOYSA-N
SMILES
Cl.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1
Infrared spectrum
Conforms
Melting point
103°C to 108°C
Appearance (Color)
White to beige
Appearance (Form)
Crystalline powder
Titration Argentometric
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text