Manganese, 99+%, powder, -40 mesh, Thermo Scientific Chemicals
Manganese, 99+%, powder, -40 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese, 99+%, powder, -40 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 317442500
Price (USD)/ Each
45.65
Online exclusive
50.60 
Save 4.95 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
45.65
Online exclusive
50.60 
Save 4.95 (10%)
Add to cart
Manganese, 99+%, powder, -40 mesh, Thermo Scientific Chemicals
Catalog number317442500
Price (USD)/ Each
45.65
Online exclusive
50.60 
Save 4.95 (10%)
-
Add to cart
Chemical Identifiers
CAS7439-96-5
IUPAC Namemanganese
Molecular FormulaMn
InChI KeyPWHULOQIROXLJO-UHFFFAOYSA-N
SMILES[Mn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey-brown to brown-black
Assay>=99.0 % (Mn)
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only