Tin, 99.8%, powder, -325 mesh, Thermo Scientific Chemicals
Tin, 99.8%, powder, -325 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin, 99.8%, powder, -325 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 317801000
Price (USD)/ Each
37.65
Online exclusive
41.70 
Save 4.05 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
37.65
Online exclusive
41.70 
Save 4.05 (10%)
Add to cart
Tin, 99.8%, powder, -325 mesh, Thermo Scientific Chemicals
Catalog number317801000
Price (USD)/ Each
37.65
Online exclusive
41.70 
Save 4.05 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Total trace metal impurities=<0.2 %
Appearance (Color)Grey
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only