Thermo Scientific Chemicals

Barium, 99+%, rods, ∽2cm diameter, Thermo Scientific Chemicals

Catalog number: 317865000
500 g, Each
Thermo Scientific Chemicals

Barium, 99+%, rods, ∽2cm diameter, Thermo Scientific Chemicals

Catalog number: 317865000
500 g, Each
Quantity
Catalog number: 317865000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-39-3
IUPAC Name
barium
Molecular Formula
Ba
InChI Key
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
SMILES
[Ba]
Assay
>=99.0 % Ba (incl. Sr)
Strontium (Sr)
=<0.8 %
Appearance (Color)
Grey
Appearance (Form)
Rods

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text