Thermo Scientific Chemicals

Cacodylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 318150500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Cacodylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 318150500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 318150500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
75-60-5
IUPAC Name
dimethylarsinic acid
Molecular Formula
C2H7AsO2
InChI Key
OGGXGZAMXPVRFZ-UHFFFAOYSA-N
SMILES
C[As](C)(O)=O
Appearance (Color)
White
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %
Appearance (Form)
Crystalline powder
Melting point
195°C to 198°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text