Thermo Scientific Chemicals

Antimony tribromide, 97%, pure, Thermo Scientific Chemicals

Catalog number: 318402500
250 g, Each
Thermo Scientific Chemicals

Antimony tribromide, 97%, pure, Thermo Scientific Chemicals

Catalog number: 318402500
250 g, Each
Quantity
Catalog number: 318402500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7789-61-9
Appearance (Color)
White to beige
Titration Argentometric
>=96.0 %
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text