Thermo Scientific Chemicals

Cobalt(II) bromide hydrate, Thermo Scientific Chemicals

Catalog number: 318545000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Cobalt(II) bromide hydrate, Thermo Scientific Chemicals

Catalog number: 318545000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 318545000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
85017-77-2
IUPAC Name
λ²-cobalt(2+) dibromide
Molecular Formula
Br2Co
InChI Key
BZRRQSJJPUGBAA-UHFFFAOYSA-L
SMILES
[Co++].[Br-].[Br-]
Appearance (Form)
Crystals
Water
24.7 to 36.5 %
Titration Argentometric
17.0 to 20.3 % (Co)
Appearance (Color)
Violet

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text