Thermo Scientific Chemicals

Gold(III) chloride, 99%, Thermo Scientific Chemicals

Catalog number: 318645000
500 mg, Glass bottle, Each
Thermo Scientific Chemicals

Gold(III) chloride, 99%, Thermo Scientific Chemicals

Catalog number: 318645000
500 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 318645000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13453-07-1
IUPAC Name
gold hydrochloridyl
Molecular Formula
AuCl
InChI Key
BRSVJNYNWNMJKC-UHFFFAOYSA-N
SMILES
[Cl].[Au]
Appearance (Form)
Crystalline powder
Appearance (Color)
Orange-red to brown to violet
Assay
>=98.5 % (ex Au)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text