Thermo Scientific Chemicals

Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals

Catalog number: 318770250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals

Catalog number: 318770250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 318770250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
51364-51-3
IUPAC Name
tris(1,5-diphenylpenta-1,4-dien-3-one) dipalladium
Molecular Formula
C51H42O3Pd2
InChI Key
CYPYTURSJDMMMP-UHFFFAOYSA-N
SMILES
[Pd].[Pd].O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1
Appearance (Color)
Purple to black
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Elemental Analysis
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text