Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
5 g
25 g
Packaging:
Glass bottle
Catalog number 318775000
Price (USD)/ Each
49.65
Online exclusive
57.90 
Save 8.25 (14%)
-
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
49.65
Online exclusive
57.90 
Save 8.25 (14%)
Add to cart
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Catalog number318775000
Price (USD)/ Each
49.65
Online exclusive
57.90 
Save 8.25 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Nametris(1,5-diphenylpenta-1,4-dien-3-one) dipalladium
Molecular FormulaC51H42O3Pd2
InChI KeyCYPYTURSJDMMMP-UHFFFAOYSA-N
SMILES[Pd].[Pd].O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1
Molecular Weight (g/mol)915.73
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Purple to black
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Elemental Analysis>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only