Chemical Identifiers
CAS69458-20-4
IUPAC NameN,N,N-trimethyl-4-{7,12,17-tris[4-(trimethylazaniumyl)phenyl]-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaen-2-yl}anilinium tetrakis(4-methylbenzene-1-sulfonate)
Molecular FormulaC84H90N8O12S4
InChI KeyAKFPMEOALZKHNR-UHFFFAOYSA-J
SMILESCC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O.C[N+](C)(C)C1=CC=C(C=C1)C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=CC=C(C=C1)[N+](C)(C)C)C1=CC=C(C=C1)[N+](C)(C)C)C1=CC=C(C=C1)[N+](C)(C)C
View more