3,4-Dichlorobenzyl isocyanate, 98%, Thermo Scientific Chemicals
3,4-Dichlorobenzyl isocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dichlorobenzyl isocyanate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 319220010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1,2-dichloro-4-(isocyanatomethyl)benzene
Molecular FormulaC8H5Cl2NO
InChI KeyWLNAHQZFPSSVTP-UHFFFAOYSA-N
SMILESClC1=CC=C(CN=C=O)C=C1Cl
Molecular Weight (g/mol)202.03
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5656 to 1.5676 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only