Thermo Scientific Chemicals

3,4-Dichlorobenzyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: 319220100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

3,4-Dichlorobenzyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: 319220100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 319220100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
19752-09-1
IUPAC Name
1,2-dichloro-4-(isocyanatomethyl)benzene
Molecular Formula
C8H5Cl2NO
InChI Key
WLNAHQZFPSSVTP-UHFFFAOYSA-N
SMILES
ClC1=CC=C(CN=C=O)C=C1Cl
Refractive index
1.5656 to 1.5676 (20°C, 589 nm)
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=97.5 %
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text