Opti-MEM™ I Reduced Serum Medium
Save up to 29% on your order of cell culture products. View promotion details >
Opti-MEM™ I Reduced Serum Medium
Gibco™

Opti-MEM™ I Reduced Serum Medium

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine SerumRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
31985062100 mL
31985070500 mL
3198508810 x 500 mL
Catalog number 31985062
Price (USD)
31.65
Online Exclusive
34.34
Save 2.69 (8%)
Each
-
Add to cart
Quantity:
100 mL
Customize this product
Price (USD)
31.65
Online Exclusive
34.34
Save 2.69 (8%)
Each
Add to cart
Ask our AI about this Product

Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology. Opti-MEM™ I medium is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ I medium can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Opti-MEM™ I modifications for a range of cell culture applications.


This Opti-MEM™ I is modified as follows:
With
• L-glutamine
• Phenol Red


The complete formulation is confidential. For more information, please contact Technical Services.

Using Opti-MEM™ I Medium
Opti-MEM™ I Reduced-Serum Medium is a unique medium that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Opti-MEM™ I medium uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineSp2, AE-1, CHO, BHK-21, and HEK
Cell TypePrimary Fibroblasts
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco™, Opti-MEM™
Product TypeOpti-MEM I
Quantity100 mL
Shelf Life18 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationHuman Origin
FormLiquid
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesGlutamine, Phenol Red
Unit SizeEach
Contents & Storage
Store at 2–8°C and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection