Thermo Scientific Chemicals

Cyclobutyl bromide, 97%, pure, Thermo Scientific Chemicals

Catalog number: 320500250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

Cyclobutyl bromide, 97%, pure, Thermo Scientific Chemicals

Catalog number: 320500250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 320500250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4399-47-7
IUPAC Name
bromocyclobutane
Molecular Formula
C4H7Br
InChI Key
KXVUSQIDCZRUKF-UHFFFAOYSA-N
SMILES
BrC1CCC1
Appearance (Form)
Liquid
Refractive index
1.4780 to 1.4810 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text