Thermo Scientific Chemicals

N,N-Dimethylformamide-d7, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D, Thermo Scientific Chemicals

Catalog number: 320730075
7.5 mL, Ampule, Each
Thermo Scientific Chemicals

N,N-Dimethylformamide-d7, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D, Thermo Scientific Chemicals

Catalog number: 320730075
7.5 mL, Ampule, Each
Quantity
Packaging
Catalog number: 320730075
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4472-41-7
IUPAC Name
N,N-di(²H₃)methyl(²H)formamide
Molecular Formula
C3H7NO
InChI Key
ZMXDDKWLCZADIW-YYWVXINBSA-N
SMILES
[2H]C(=O)N(C([2H])([2H])[2H])C([2H])([2H])[2H]
Appearance (Form)
Liquid
Water
=<0.1 % (Coulometric)
D-Enrichment
>=99.4 %
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text