Tetravinyl tin, 95%, Thermo Scientific Chemicals
Tetravinyl tin, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetravinyl tin, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number 321080250
Price (USD)/ Each
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1112-56-7
IUPAC Nametetraethenylstannane
Molecular FormulaC8H12Sn
InChI KeyMZIYQMVHASXABC-UHFFFAOYSA-N
SMILESC=C[Sn](C=C)(C=C)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5000 to 1.5030 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only