Thermo Scientific™

Pierce™ ECL Plus Western Blotting Substrate

Catalog number: 32132
Have Questions?
Thermo Scientific™

Pierce™ ECL Plus Western Blotting Substrate

Catalog number: 32132
Have Questions?
Catalog Number
32132
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
300 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
32132100 mL
-
3213425 mL
-
32132X3300 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce ECL Plus Substrate is an acridan-based chemiluminescent and chemifluorescent HRP substrate for western blot detection using X-ray film or CCD- or laser-based imagers. This product is sold as Pierce ECL 2 Substrate (Cat. #PI80196X3) through Fisher Scientific and other channels.

Features of ECL Plus Substrate:

Easy to use—can be substituted for the discontinued GE ECL Plus Substrate without any re-optimization
Higher sensitivity—detect targets down to the low-picogram level
Longer signal duration—sustained light output for as long as 5 hours
More imaging options—X-ray, CCD or laser-based imagers
More affordable—high quality and performance at a lower price than other suppliers' similar substrates

Pierce ECL Plus Substrate generates acridinium esters when it reacts with HRP. As these ester intermediates react with peroxide, they produce a strong and sustained chemiluminescent signal that can be captured by X-ray film CCD imaging. The substrate also produces a robust fluorescent signal that can be captured with fluorescence imagers. Pierce ECL Plus Substrate enables the detection of low-picogram amounts of target protein on nitrocellulose or PVDF membrane when probed with appropriate primary and secondary antibody concentrations. Its broad dynamic range, high sensitivity and long-lasting signal make Pierce ECL Plus Substrate an excellent choice for western blot analysis.
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line
Pierce™
Recommended Antibody Concentrations
2°: 1:25-200K (5-40ng/mL), 1°: 1:1K (0.05- 1µg/mL)
Signal Duration
5 hr
Substrate Properties
Chemical Substrate
Substrate Type
HRP (Horseradish Peroxidase) Substrate
Detection Method
Chemiluminescent and Fluorescent
Dimensions (L x W x H)
8.37 x 4.37 x 4.37 in. (21.2 x 11 x 11cm)
Sensitivity
Low picogram to mid-femtogram

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text