1,2-Bis(dichlorophosphino)ethane, 97%, Thermo Scientific Chemicals
1,2-Bis(dichlorophosphino)ethane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Bis(dichlorophosphino)ethane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 321470050
Price (USD)/ Each
215.65
Online exclusive
254.00 
Save 38.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
215.65
Online exclusive
254.00 
Save 38.35 (15%)
Add to cart
1,2-Bis(dichlorophosphino)ethane, 97%, Thermo Scientific Chemicals
Catalog number321470050
Price (USD)/ Each
215.65
Online exclusive
254.00 
Save 38.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedichloro[2-(dichlorophosphanyl)ethyl]phosphane
Molecular FormulaC2H4Cl4P2
InChI KeySBWAJHLQMFBNIN-UHFFFAOYSA-N
SMILESClP(Cl)CCP(Cl)Cl
Molecular Weight (g/mol)231.80
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5875 to 1.5935 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
NMR>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only