(Bromomethyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals
(Bromomethyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(Bromomethyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 321480050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1034-49-7
IUPAC Name(bromomethyl)triphenylphosphanium bromide
Molecular FormulaC19H17Br2P
InChI KeyYFTMLUSIDVFTKU-UHFFFAOYSA-M
SMILES[Br-].BrC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=97.5 %
Appearance (Color)White to light beige to pink
Infrared spectrumConforms
Appearance (Form)Powder or crystals
Melting point232°C to 242°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only