Thermo Scientific Chemicals

(Bromomethyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: 321480250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

(Bromomethyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: 321480250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 321480250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1034-49-7
IUPAC Name
(bromomethyl)triphenylphosphanium bromide
Molecular Formula
C19H17Br2P
InChI Key
YFTMLUSIDVFTKU-UHFFFAOYSA-M
SMILES
[Br-].BrC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Titration Argentometric
>=97.5 %
Appearance (Color)
White to light beige to pink
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals
Melting point
232°C to 242°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text