chlorodiisopropylphosphine, 96%, Thermo Scientific Chemicals
chlorodiisopropylphosphine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

chlorodiisopropylphosphine, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 321550050
Price (USD)/ Each
73.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
86.00 
Save 12.35 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
73.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
86.00 
Save 12.35 (14%)
Add to cart
chlorodiisopropylphosphine, 96%, Thermo Scientific Chemicals
Catalog number321550050
Price (USD)/ Each
73.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
86.00 
Save 12.35 (14%)
-
Add to cart
Chemical Identifiers
CAS40244-90-4
IUPAC Namechlorobis(propan-2-yl)phosphane
Molecular FormulaC6H14ClP
InChI KeyJZPDBTOWHLZQFC-UHFFFAOYSA-N
SMILESCC(C)P(Cl)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow
Titration Argentometric>=95 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4740 to 1.4760 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only