Thermo Scientific Chemicals

Bis(diisopropylamino)chlorophosphine, Thermo Scientific Chemicals

Catalog number: 321560050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Bis(diisopropylamino)chlorophosphine, Thermo Scientific Chemicals

Catalog number: 321560050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 321560050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56183-63-2
Appearance (Form)
Powder or crystals and/or chunks
Melting point
100°C to 110°C
Appearance (Color)
White to off-white to yellow
Infrared spectrum
Conforms
Additional info
May contain varying amount of diisopropylamine.HCl

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text