Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ Nalgene Square Amber PETG Media Bottles include HDPE closures. Translucent amber bottles meet USP light-resistant requirements and are ideal for medias, buffers and sera.
Capacity (Metric) | 60 mL |
Capacity (English) | 2 oz. |
Color | Amber |
Diameter (English) | 1.6 in. |
Diameter (English) Inner Neck | 0.71 in. |
Diameter (Metric) Inner Neck | 18 mm |
Material | PETG |
No. per Case | 200 |
No. per Pack | 40 |
Sterility | Non-sterile |
Certifications/Compliance | PETG Media Bottles and HDPE closure materials meet current biological testing standards including USP Class VI. |
Closure Included | Yes |
Graduated | Yes |
Mouth | Wide |
Shape | Square |
Height (Metric) | 82 mm |
Product Type | Media Bottle |
Height (English) | 3.2 in. |
Closure Material | HDPE |
Closure Size | 24 mm |
Diameter (Metric) | 41 mm |
Unit Size | Case of 200 |
Capacity (Metric) | 30 mL |
Capacity (English) | 1 oz. |
Color | Amber |
Diameter (English) | 1.5 in. |
Diameter (English) Inner Neck | 0.55 in. |
Diameter (Metric) Inner Neck | 14 mm |
Material | PETG |
No. per Case | 280 |
No. per Pack | 40 |
Sterility | Non-sterile |
Certifications/Compliance | PETG Media Bottles and HDPE closure materials meet current biological testing standards including USP Class VI. |
Closure Included | Yes |
Graduated | Yes |
Mouth | Wide |
Shape | Square |
Height (Metric) | 64 mm |
Product Type | Media Bottle |
Height (English) | 2.5 in. |
Closure Material | HDPE |
Closure Size | 20 mm |
Diameter (Metric) | 38 mm |
Unit Size | Case of 280 |
Capacity (Metric) | 125 mL |
Capacity (English) | 4.2 oz. |
Color | Amber |
Diameter (English) | 2.1 in. |
Diameter (English) Inner Neck | 1.1 in. |
Diameter (Metric) Inner Neck | 28 mm |
Material | PETG |
No. per Case | 96 |
No. per Pack | 24 |
Sterility | Non-sterile |
Certifications/Compliance | PETG Media Bottles and HDPE closure materials meet current biological testing standards including USP Class VI. |
Closure Included | Yes |
Graduated | Yes |
Mouth | Wide |
Shape | Square |
Height (Metric) | 110 mm |
Product Type | Media Bottle |
Height (English) | 4.3 in. |
Closure Material | HDPE |
Closure Size | 38-430 mm |
Diameter (Metric) | 54 mm |
Unit Size | Case of 96 |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | dimClosureSize | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
322021-0060 | Case of 200 | 60 mL | 356.00 Online price 565.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
322021-0030 | Case of 280 | 30 mL | 425.00 Online price 674.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
322021-0125 | Case of 96 | 125 mL | 331.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Ordering Information:
Products are shrink-wrapped into trays, then packed in double PE lined cartons for cleanroom applications.
Recommended for:
For storage and shipping of light sensitive liquid media, buffers, and sera
Note:
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.