Search by lot number or partial lot number
Protect against leakage, breakage, and contamination using Thermo Scientific™ Nalgene Nonsterile Serum Vials. Lighter than glass. Use with lined continuous thread closures (available separately).
Description | Serum Vials |
Diameter (Metric) Inner | 11 mm |
Color | Clear |
Diameter (Metric) Outer | 24 mm |
Material | PETG |
Sterility | Non-sterile |
Volume (Metric) | 10 mL |
No. per Case | 1260 |
No. per Pack | 252 |
Volume (English) | 1/3 oz. |
Product Line | Nalgene |
Volume (Metric) Total | 10 mL |
Closure Type | Screw Cap (Not Included) |
Height (Metric) | 54 mm |
Quantity | 252 Pk., 1260 Cases |
Certifications/Compliance | ISO 13485:2003 |
Type | Serum Vial |
Closure Size | 20-415 |
For Use With (Equipment) | Lined Continuous Thread Closure |
Unit Size | Case of 1260 |
Description | Serum Vials |
Diameter (Metric) Inner | 11 mm |
Color | Clear |
Diameter (Metric) Outer | 22 mm |
Material | PETG |
Sterility | Non-sterile |
Volume (Metric) | 5 mL |
No. per Case | 1656 |
No. per Pack | 276 |
Volume (English) | 1/6 oz. |
Product Line | Nalgene |
Volume (Metric) Total | 5 mL |
Closure Type | Screw Cap (Not Included) |
Height (Metric) | 44 mm |
Quantity | 276 Pk. , 1656 Cases |
Certifications/Compliance | ISO 13485:2003 |
Type | Serum Vial |
Closure Size | 20-415 |
For Use With (Equipment) | Lined Continuous Thread Closure |
Unit Size | Case of 1656 |
Description | Serum Vials |
Diameter (Metric) Inner | 11 mm |
Color | Clear |
Diameter (Metric) Outer | 30 mm |
Material | PETG |
Sterility | Non-sterile |
Volume (Metric) | 20 mL |
No. per Case | 612 |
No. per Pack | 153 |
Volume (English) | 2/3 oz. |
Product Line | Nalgene |
Volume (Metric) Total | 20 mL |
Closure Type | Continuous-thread Closure (Sold Separately) |
Height (Metric) | 64 mm |
Quantity | 153 Pk., 612 Cases |
Certifications/Compliance | ISO 13485:2003 |
Type | Serum Vial |
Closure Size | 20-415 |
For Use With (Equipment) | Lined Continuous Thread Closure |
Unit Size | Case of 612 |
Catalog Number | Specifications | Unit Size | Volume (Metric) | Closure Size | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
322032-0010 | Case of 1260 | 10 mL | 20-415 | 391.00 Online price 620.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
322032-0005 | Case of 1656 | 5 mL | 20-415 | 579.20 Online price 724.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
322032-0020 | Case of 612 | 20 mL | 20-415 | 232.00 Online price 367.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Ordering Information:
Serum Vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 312158).